1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether ≥98%

Supplier: Matrix Scientific

004978-1G 004978-5G
101788-106EA 36.92 USD
101788-106 101788-108
1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether ≥98%
1,1-Dichloro-2,2,2-trifluoroethylchlorodifluoromethyl ether

Hazard: Irritant

Formula: C₃Cl₃F₅O
MW: 253.38 g/mol
Boiling Pt: 74.5°C
MDL Number: MFCD01320777
CAS Number: 32778-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR