AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride) ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride

004872-250MG 004872-1G
101787-936EA 103.08 USD
101787-936 101787-938
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride) ≥98%
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulphonylfluoride hydrochloride)

Hazard: Irritant/Refrigerate

Formula: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
Melting Pt: 184 °C (dec.)
MDL Number: MFCD00132962
CAS Number: 30827-99-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR