6-Chloro-3,5-dibromo-2,2,3,4,4,5,6,6-octafluorohexanenitrile ≥97%

Supplier: Matrix Scientific

Synonyms: Perfluoro-6-chloro-3,5-dibromohexanenitrile

004556-1G 004556-5G
101787-470EA 73.85 USD
101787-470 101787-472
6-Chloro-3,5-dibromo-2,2,3,4,4,5,6,6-octafluorohexanenitrile ≥97%
6-Chloro-3,5-dibromo-2,2,3,4,4,5,6,6-octafluorohexanenitrile

Hazard: Irritant

MW: 433.32 g/mol
MDL Number: MFCD08461581
CAS Number: 240800-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR