Ethyl-2-fluoroacetoacetate

Supplier: Matrix Scientific

004384-5G 004384-25G
101787-136EA 33.85 USD
101787-136 101787-138
Ethyl-2-fluoroacetoacetate
Ethyl-2-fluoroacetoacetate

Hazard: Irritant

Formula: C₆H₉FO₃
Boiling Pt: 183 °C
Density: 1.181
Flash Pt: 90 °C
MDL Number: MFCD00215830
CAS Number: 1522-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR