2-(Tributylstannyl)pyrazine ≥90%

Supplier: Matrix Scientific

004350-1G 004350-5G
101787-102EA 130.77 USD
101787-102 101787-104
2-(Tributylstannyl)pyrazine ≥90%
2-(Tributylstannyl)pyrazine

Balance is non-aromatic tri and tetraalkyltins


Hazard: Toxic/Refrigerate

Formula: C₁₆H₃₀N₂Sn
Boiling Pt: 130°C/1mm
Density: 1.171
Flash Pt: 110°C
MDL Number: MFCD01319077
CAS Number: 205371-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR