4-(Dimethylamino)-3-nitrobenzotrifluoride ≥97%

Supplier: Matrix Scientific

004276-1G 004276-5G
101787-046EA 70.77 USD
101787-046 101787-048
4-(Dimethylamino)-3-nitrobenzotrifluoride ≥97%
4-(Dimethylamino)-3-nitrobenzotrifluoride

Hazard: Irritant/Irritant-Harmful

Formula: C₉H₉F₃N₂O₂
MDL Number: MFCD00236670
CAS Number: 40700-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR