Dibenzothiophene-4-boronic acid

Supplier: Matrix Scientific

Synonyms: 4-Dibenzothiopheneboronic acid

004206-5G 004206-25G
101786-956EA 35.38 USD
101786-956 101786-958
Dibenzothiophene-4-boronic acid
Dibenzothiophene-4-boronic acid

Hazard: Irritant/Store in Freezer

Formula: C₁₂H₉BO₂S
MW: 228.08 g/mol
Melting Pt: 337…339 °C
MDL Number: MFCD01318182
CAS Number: 108847-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR