Phenoxathiin-4-boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: Phenoxathiin-4-ylboronic acid

004158-1G 004158-5G
101786-876EA 158.46 USD
101786-876 102550-838
Phenoxathiin-4-boronic acid ≥97%
Phenoxathiin-4-boronic acid

Hazard: Irritant/Refrigerate

Formula: C₁₂H₉BO₃S
MW: 244.08 g/mol
Melting Pt: 156…157 °C
MDL Number: MFCD01605731
CAS Number: 100124-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR