3,5-Dimethylisoxazole-4-boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (dimethyl-1,2-oxazol-4-yl)boronic acid

004078-5G 004078-25G
101786-800EA 66.15 USD
101786-800 101786-802
3,5-Dimethylisoxazole-4-boronic acid ≥97%
3,5-Dimethylisoxazole-4-boronic acid

Hazard: Irritant/Refrigerate

Formula: C₅H₈BNO₃
MW: 140.93 g/mol
Melting Pt: 106-108°C
MDL Number: MFCD02677945
CAS Number: 16114-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR