Dimethyl hexadecafluorosebacate ≥96%

Supplier: Matrix Scientific

Synonyms: Dimethyl perfluorosebacate

003936-5G 003936-25G
101786-478EA 35.38 USD
101786-478 101786-480
Dimethyl hexadecafluorosebacate ≥96%
Dimethyl hexadecafluorosebacate

Hazard: Irritant

Formula: C₁₂H₆F₁₆O₄
Boiling Pt: 144…146 °C
MDL Number: MFCD00236604
CAS Number: 4590-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR