4-Fluoro-DL-phenylglycine

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-DL-α-phenylglycine, 4-Fluorophenylglycine, H-DL-Phg(4-F)-OH

003921-5G 003921-25G
101786-454EA 69.23 USD
101786-454 101786-456
4-Fluoro-DL-phenylglycine
4-Fluoro-DL-phenylglycine

Hazard: Irritant

Formula: C₈H₈FNO₂
MW: 169.155 g/mol
Melting Pt: 270…271 °C
MDL Number: MFCD00066444
CAS Number: 7292-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR