2,4,5-Trifluorocinnamic acid

Supplier: Matrix Scientific

003691-1G 003691-5G
101786-084EA 35.38 USD
101786-084 101786-086
2,4,5-Trifluorocinnamic acid
2,4,5-Trifluorocinnamic acid

Hazard: Irritant

Formula: C₉H₅F₃O₂
Melting Pt: 184…188 °C
MDL Number: MFCD00061211
CAS Number: 247170-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR