2,3,6-Trifluorocinnamic acid

Supplier: Matrix Scientific

Synonyms: 3-(2,3,6-Trifluorophenyl)acrylic acid

003690-1G 003690-5G
101786-080EA 47.69 USD
101786-080 101786-082
2,3,6-Trifluorocinnamic acid
2,3,6-Trifluorocinnamic acid

Hazard: Irritant

Formula: C₉H₅F₃O₂
MW: 202.13 g/mol
MDL Number: MFCD00061213
CAS Number: 230295-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR