2,3,5-Trifluorocinnamic acid

Supplier: Matrix Scientific

003689-1G 003689-5G
101786-076EA 60 USD
101786-076 101786-078
2,3,5-Trifluorocinnamic acid
2,3,5-Trifluorocinnamic acid

Hazard: Irritant

Formula: C₉H₅F₃O₂
MW: 202.13 g/mol
CAS Number: 237761-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR