2,3,6-Trifluoroanisole

Supplier: Matrix Scientific

003619-1G 003619-5G
101785-938EA 50.77 USD
101785-938 101785-940
2,3,6-Trifluoroanisole
2,3,6-Trifluoroanisole

Hazard: Flammable

Formula: C₇H₅F₃O
Boiling Pt: 146 °C
Flash Pt: 45 °C
MDL Number: MFCD01631501
CAS Number: 4920-34-7
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR