1H,1H-Perfluoro-n-decyl methacrylate ≥97%

Supplier: Matrix Scientific

003597-5G 003597-25G
101785-910EA 89.23 USD
101785-910 101785-912
1H,1H-Perfluoro-n-decyl methacrylate ≥97%
1H,1H-Perfluoro-n-decyl methacrylate

Hazard: Irritant/Refrigerate/Lachrymator

Formula: C₁₄H₇F₁₉O₂
MW: 568.18 g/mol
Boiling Pt: 77 °C (1 mmHg)
MDL Number: MFCD02183511
CAS Number: 23069-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR