2,3,4,5-Tetrafluorobenzamide
Supplier: Matrix Scientific
003453-1G
003453-5G
101785-722EA
35.38
USD
101785-722
101785-724
2,3,4,5-Tetrafluorobenzamide
2,3,4,5-Tetrafluorobenzamide
Hazard: Irritant
Formula:
C₇H₃F₄NO |
MDL Number:
MFCD01631457 CAS Number: 16582-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...