4-(Methylsulfonyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid

003366-5G 003366-25G
101785-620EA 44.62 USD
101785-620 102663-554
4-(Methylsulfonyl)phenylboronic acid ≥97%
4-(Methylsulphonyl)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₉BO₄S
MW: 200.02 g/mol
MDL Number: MFCD01630820
CAS Number: 149104-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR