5-Indoleboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid

003035-1G 003035-5G
101785-118EA 41.54 USD
101785-118 101785-120
5-Indoleboronic acid ≥97%
5-Indoleboronic acid

Hazard: Irritant/Refrigerate

Formula: C₈H₈BNO₂
MW: 160.97 g/mol
Melting Pt: 173…174 °C
MDL Number: MFCD01319013
CAS Number: 144104-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR