2-Chloro-N-(3-fluorophenyl)acetamide ≥97%

Supplier: Matrix Scientific

002926-5G 002926-25G 002926-100G
101784-926EA 13.85 USD
101784-926 101784-928 101784-930
2-Chloro-N-(3-fluorophenyl)acetamide ≥97%
2-Chloro-N-(3-fluorophenyl)acetamide

Hazard: Irritant

Formula: C₈H₇ClFNO
Melting Pt: 122 °C
MDL Number: MFCD01034343
CAS Number: 350-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR