3',4',5'-Trifluoropropiophenone ≥97%

Supplier: Matrix Scientific

002888-1G 002888-5G
101784-860EA 67.69 USD
101784-860 101784-862
3',4',5'-Trifluoropropiophenone ≥97%
3',4',5'-Trifluoropropiophenone

Hazard: Irritant

Formula: C₉H₇F₃O
MW: 188.15
CAS Number: 220227-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR