2',4',5'-Trifluoropropiophenone ≥97%

Supplier: Matrix Scientific

002887-1G 002887-5G
101784-856EA 46.15 USD
101784-856 101784-858
2',4',5'-Trifluoropropiophenone ≥97%
2',4',5'-Trifluoropropiophenone

Hazard: Irritant

Formula: C₉H₇F₃O
MW: 188.15
MDL Number: MFCD00061223
CAS Number: 220228-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR