2',3',6'-Trifluoropropiophenone ≥97%

Supplier: Matrix Scientific

002886-1G 002886-5G
101784-852EA 52.31 USD
101784-852 101784-854
2',3',6'-Trifluoropropiophenone ≥97%
2',3',6'-Trifluoropropiophenone

Hazard: Irritant

Formula: C₉H₇F₃O
MW: 188.15
MDL Number: MFCD00061222
CAS Number: 243666-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR