2',3',5'-Trifluoropropiophenone ≥97%
Supplier: Matrix Scientific
002885-1G
002885-5G
101784-848EA
73.85
USD
101784-848
101784-850
2',3',5'-Trifluoropropiophenone ≥97%
2',3',5'-Trifluoropropiophenone
Hazard: Irritant
Formula:
C₉H₇F₃O |
MDL Number:
MFCD00083545 CAS Number: 243666-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...