N-Boc-4-(trifluoromethyl)-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: N-(tert-Butoxycarbonyl)-4-trifluoromethyl-L-phenylalanine, Boc-Phe(4-CF3)-OH, N-Boc-4-trifluoromethyl-L-phenylalanine

002881-250MG 002881-1G
101784-836EA 61.54 USD
101784-836 101784-838
N-Boc-4-(trifluoromethyl)-L-phenylalanine
N-Boc-4-(trifluoromethyl)-L-phenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈F₃NO₄
MW: 333.31 g/mol
MDL Number: MFCD00792407
CAS Number: 114873-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR