2-Amino-3-thiophenecarbonitrile

Supplier: Matrix Scientific

Synonyms: 2-Aminothiophene-3-carbonitrile

029256-25G 029256-100G
101827-788EA 75.38 USD
101827-788 101827-790
2-Amino-3-thiophenecarbonitrile
2-Amino-3-thiophenecarbonitrile

Hazard: Irritant

Formula: C₅H₄N₂S
MW: 124.16 g/mol
MDL Number: MFCD00706298
CAS Number: 4651-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR