1-(Methylsulfonyl)piperazine

Supplier: Matrix Scientific

Synonyms: 1-Methanesulphonylpiperazine

028784-5G 028784-25G
101826-842EA 41.54 USD
101826-842 101826-844
1-(Methylsulfonyl)piperazine
1-(Methylsulphonyl)piperazine

Hazard: Irritant/Store in Freezer

Formula: C₅H₁₂N₂O₂S
MW: 164.23 g/mol
MDL Number: MFCD02153225
CAS Number: 55276-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR