4-Methylcyclohexylamine (cis- and trans- mixture)

Supplier: Matrix Scientific

028770-25G 028770-100G
101826-812EA 110.77 USD
101826-812 101826-814
4-Methylcyclohexylamine (cis- and trans- mixture)
4-Methylcyclohexylamine (cis- and trans- mixture)

Hazard: Irritant

Formula: C₇H₁₅N
MDL Number: MFCD00001495
CAS Number: 6321-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR