3-Methyl-1H-pyrazole-5-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 5-Methyl-1H-pyrazole-3-carbaldehyde

016461-500MG 016461-2.5G
101804-774EA 221.54 USD
101804-774 101804-776
3-Methyl-1H-pyrazole-5-carbaldehyde
3-Methyl-1H-pyrazole-5-carbaldehyde

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00159629
CAS Number: 3273-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR