1-Thiophen-2-ylethylamine ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Thiophen-2-yl)ethanamine, 2-(1-Aminoethyl)thiophene

010302-250MG 010302-1G
101798-262EA 80 USD
101798-262 101798-264
1-Thiophen-2-ylethylamine ≥97%
1-Thiophen-2-ylethylamine

Hazard: Corrosive/Air Sensitive

Formula: C₆H₉NS
MW: 127.21 g/mol
Boiling Pt: 83-84°C/16 Torr
Density: 1.0931/15°C
MDL Number: MFCD02734311
CAS Number: 6309-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR