6-Bromoquinoline

Supplier: Matrix Scientific

010062-5G 010062-25G
101797-802EA 20 USD
101797-802 101797-804
6-Bromoquinoline
6-Bromoquinoline

Hazard: Irritant

Formula: C₉H₆BrN
Boiling Pt: 281 °C
Melting Pt: 24 °C
MDL Number: MFCD00024023
CAS Number: 5332-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR