3,4-Diaminofurazan ≥97%

Supplier: Matrix Scientific

010059-1G 010059-5G
101797-794EA 36.92 USD
101797-794 101797-796
3,4-Diaminofurazan ≥97%
3,4-Diaminofurazan

Hazard: Irritant

Formula: C₂H₄N₄O
Melting Pt: 179…180 °C
MDL Number: MFCD00138084
CAS Number: 17220-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR