5-Amino-1,3-dihydro-2H-benzimidazol-2-one ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-benzimidazolinone

010022-25G 010022-100G
101797-718EA 16.92 USD
101797-718 101797-720
5-Amino-1,3-dihydro-2H-benzimidazol-2-one ≥97%
5-Amino-1,3-dihydro-2H-benzimidazol-2-one

Hazard: Irritant

Formula: C₇H₇N₃O
MW: 149.1500 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00053555
CAS Number: 95-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR