4-Bromo-7-fluoroindole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-7-fluoro-1H-indole

009817-500MG 009817-2.5G
101797-318EA 380 USD
101797-318 101797-320
4-Bromo-7-fluoroindole ≥95%
4-Bromo-7-fluoroindole

Hazard: Irritant

Formula: C₈H₅BrFN
MW: 214.04 g/mol
Melting Pt: 24…25 °C
MDL Number: MFCD03095318
CAS Number: 883500-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR