4-Methyl-3-nitrophenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Methyl-3-nitrophenyl)boronic acid, 4-Methyl-3-nitrobenzeneboronic acid

009800-5G 009800-25G
101797-292EA 75.38 USD
101797-292 101797-294
4-Methyl-3-nitrophenylboronic acid ≥97%
4-Methyl-3-nitrophenylboronic acid

Hazard: Irritant

Formula: C₇H₈BNO₄
MW: 180.96 g/mol
MDL Number: MFCD00191550
CAS Number: 80500-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR