4-Carboxyphenylboronic acid pinacol ester ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
009765-1G
009765-5G
009765-25G
101797-200EA
15.38
USD
101797-200
101797-202
101797-204
4-Carboxyphenylboronic acid pinacol ester ≥97%
4-Carboxyphenylboronic acid pinacol ester
Hazard: Irritant/Refrigerate
Formula:
C₁₃H₁₇BO₄ Melting Pt: 227-231°C |
MDL Number:
MFCD01863710 CAS Number: 180516-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...