5-Bromo-3-methylindole ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-methyl-1H-indole

009761-1G 009761-5G
101797-188EA 113.85 USD
101797-188 101797-190
5-Bromo-3-methylindole ≥97%
5-Bromo-3-methylindole

Hazard: Irritant/Refrigerate

Formula: C₉H₈BrN
MW: 210.08 g/mol
MDL Number: MFCD04038746
CAS Number: 10075-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR