5-Fluoro-m-toluic acid ≥98%

Supplier: Matrix Scientific

009530-1G 009530-5G
101797-146EA 16.92 USD
101797-146 101797-148
5-Fluoro-m-toluic acid ≥98%
5-Fluoro-m-toluic acid

Hazard: Irritant

Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD03094313
CAS Number: 518070-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR