(4-Chloro-3-fluorophenyl)acetic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
2-(4-Chloro-3-fluorophenyl)acetic acid
009488-1G
009488-5G
009488-25G
101797-130EA
21.54
USD
101797-130
101797-132
101797-134
(4-Chloro-3-fluorophenyl)acetic acid ≥98%
(4-Chloro-3-fluorophenyl)acetic acid
Hazard: Irritant
Formula:
(C₆H₃ClF)CH₂COOH MW: 188.59 g/mol |
MDL Number:
MFCD04115861 CAS Number: 883500-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...