2-Bromo-4-fluorophenylacetonitrile ≥95%

Supplier: Matrix Scientific

009435-5G 009435-25G
101797-108EA 20 USD
101797-108 101797-110
2-Bromo-4-fluorophenylacetonitrile ≥95%
2-Bromo-4-fluorophenylacetonitrile

Hazard: Irritant

Formula: C₈H₅BrFN
MDL Number: MFCD04115829
CAS Number: 61150-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR