ω-Chloro-2,5-difluoroacetophenone ≥96%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-2',5'-Difluoroacetophenone

009427-5G 009427-25G
101797-072EA 30.77 USD
101797-072 101797-074
ω-Chloro-2,5-difluoroacetophenone ≥96%
ω-Chloro-2,5-difluoroacetophenone

Hazard: Irritant/Lachrymator

Formula: C₈H₅ClF₂O
MW: 190.5745 g/mol
Melting Pt: 49…50 °C
MDL Number: MFCD04115827
CAS Number: 60468-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR