3,5-Bis(methylsulfonyl)benzoic acid

Supplier: Matrix Scientific

009384-250MG 009384-1G
101796-988EA 590.77 USD
101796-988 101796-990
3,5-Bis(methylsulfonyl)benzoic acid
3,5-Bis(methylsulfonyl)benzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₆S₂
Melting Pt: 283…285 °C
MDL Number: MFCD04037979
CAS Number: 90536-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR