2-(1,3-Benzoxazol-2-yl)-1-[4-(trifluoromethyl)phenyl]ethanone
Supplier: Matrix Scientific
009296-1G
009296-5G
101796-806EA
127.69
USD
101796-806
101796-808
2-(1,3-Benzoxazol-2-yl)-1-[4-(trifluoromethyl)phenyl]ethanone
2-(1,3-Benzoxazol-2-yl)-1-[4-(trifluoromethyl)phenyl]ethanone
Hazard: Irritant
Formula:
C₁₆H₁₀F₃NO₂ MW: 305.26 g/mol |
MDL Number:
MFCD04037912 CAS Number: 849021-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...