3-Bromo-4-fluorophenylacetic acid ≥98%

Supplier: Matrix Scientific

009244-5G 009244-25G
101796-704EA 26.15 USD
101796-704 101796-706
3-Bromo-4-fluorophenylacetic acid ≥98%
3-Bromo-4-fluorophenylacetic acid

Hazard: Irritant

Formula: C₈H₆BrFO₂
MW: 233.04 g/mol
MDL Number: MFCD04038373
CAS Number: 194019-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR