4-(Trifluoromethyl)phenyl acetate ≥97%

Supplier: Matrix Scientific

009206-1G 009206-5G
101796-636EA 346.15 USD
101796-636 101796-638
4-(Trifluoromethyl)phenyl acetate ≥97%
4-(Trifluoromethyl)phenyl acetate

Hazard: Irritant

Formula: C₉H₇F₃O₂
MDL Number: MFCD00829056
CAS Number: 78950-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR