1,1,1-Trifluoroacetone oxime ≥97%

Supplier: Matrix Scientific

009183-1G 009183-5G
101796-588EA 80 USD
101796-588 101796-590
1,1,1-Trifluoroacetone oxime ≥97%
1,1,1-Trifluoroacetone oxime

Hazard: Irritant

Formula: C₃H₄F₃NO
MDL Number: MFCD06248847
CAS Number: 431-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR