Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ≥97%

Supplier: Matrix Scientific

009157-1G 009157-5G
101796-524EA 106.15 USD
101796-524 101796-526
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ≥97%
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Hazard: Irritant/Stored under Argon

Formula: C₃₂H₁₂BF₂₄Na
Melting Pt: 300…306 °C
MDL Number: MFCD00043323
CAS Number: 79060-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR