Potassium-2,2,3,3-tetrafluoropropionate ≥97%

Supplier: Matrix Scientific

009148-5G 009148-25G
101796-492EA 1461.54 USD
101796-492 101796-494
Potassium-2,2,3,3-tetrafluoropropionate ≥97%
Potassium-2,2,3,3-tetrafluoropropionate

Hazard: Irritant

Formula: C₃HF₄KO₂
MW: 184.14 g/mol
MDL Number: MFCD03094125
CAS Number: 71592-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR