Perfluoro-3,6,9-trioxaundecane-1,11-dioic acid ≥98%

Supplier: Matrix Scientific

009141-5G 009141-25G
101796-466EA 181.54 USD
101796-466 101796-468
Perfluoro-3,6,9-trioxaundecane-1,11-dioic acid ≥98%
Perfluoro-3,6,9-trioxaundecane-1,11-dioic acid

Hazard: Corrosive

Formula: C₈H₂F₁₂O₇
Melting Pt: 60…62 °C
MDL Number: MFCD03094427
CAS Number: 55621-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR