1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluorodocosane

Supplier: Matrix Scientific

009128-1G 009128-5G
101796-432EA 300 USD
101796-432 101796-434
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluorodocosane
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluorodocosane

Hazard: Irritant

Formula: C₂₂H₂₉F₁₇
Boiling Pt: 526 °C
Melting Pt: 90 °C
MDL Number: MFCD04038348
CAS Number: 133310-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR